4-Methoxysalicylaldehyde ≥98%

Synonyms: 2-Hydroxy-4-methoxybenzaldehyde

160695-25G 160695-5G
220039-105EA 619.23 USD
220039-105 220039-106
4-Methoxysalicylaldehyde ≥98%
4-Methoxysalicylaldehyde
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 124 °C (12 mmHg)
Melting Pt: 42…43 °C
Density: 1.2065 g/cm³ (51 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003327
CAS Number: 673-22-3
EINECS: 211-604-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR