3',4'-Dichloroacetophenone ≥99%

Synonyms: 1-(3,4-dichlorophenyl)ethanone

160520-25G 160520-100G
220038-443EA 28 USD
220038-443 220038-442
3',4'-Dichloroacetophenone ≥99%
3',4'-Dichloroacetophenone
Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 44…46 °C
MDL Number: MFCD00000553
CAS Number: 2642-63-9
EINECS: 220-146-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR