9-Fluorenylmethanol

Synonyms: (Fluoren-9-yl)methanol, 9-Fluorenmethanol, 9-Fluorenemethanol

160504-50G 160504-10G
220038-438EA 358.62 USD
220038-438 220038-437
9-Fluorenylmethanol
9-Fluorenylmethanol
Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 107 °C
MDL Number: MFCD00001139
CAS Number: 24324-17-2
EINECS: 246-167-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR