2-Bromothiazole

160474-5G 160474-25G
220038-436EA 56.86 USD
220038-436 220038-435
2-Bromothiazole
2-Bromothiazole
Formula: C₃H₂BrNS
Boiling Pt: 171 °C (1013 hPa)
Density: 1.82 g/cm³ (20 °C)
MDL Number: MFCD00005316
CAS Number: 3034-53-5
EINECS: 221-229-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR