Dibenzyl malonate

160407-10G 160407-50G
220039-099EA 79.26 USD
220039-099 220039-100
Dibenzyl malonate
Dibenzyl malonate
Formula: C₁₇H₁₆O₄
Boiling Pt: 188 °C (1013 hPa)
Density: 1.137 g/cm³ (20 °C)
MDL Number: MFCD00004779
CAS Number: 15014-25-2
EINECS: 239-099-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR