Phthalimide potassium salt

Synonyms: N-Potassium phthalimide, Potassium 1,3-dioxoisoindolin-2-ide, Potassium phthalimide

160385-500G 160385-5G 160385-100G
220039-091EA 108.77 USD
220039-091 220039-092 220039-090
Phthalimide potassium salt
Phthalimide potassium salt
Formula: C₈H₄KNO₂
MW: 185.22 g/mol
Melting Pt: ? 300 °C
Density: 1.634 g/cm³ (20 °C)
MDL Number: MFCD00005887
CAS Number: 1074-82-4
EINECS: 214-046-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR