4,4'-Bis(diethylamino)benzophenone

Warning

Synonyms: N,N,N',N'-Tetraethyl-4,4'-diaminobenzophenone, 4,4-Bis(diethylamino) benzophenone

160326-25G 160326-100G
220039-080EA 151.69 USD
220039-080 220039-079
4,4'-Bis(diethylamino)benzophenone
4,4'-Bis(diethylamino)benzophenone
Formula: C₂₁H₂₈N₂O
MW: 324.47 g/mol
Boiling Pt: 420 °C (1013 hPa)
Melting Pt: 96.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00009044
CAS Number: 90-93-7
EINECS: 202-025-4
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR