3-Methoxybenzylamine ≥98%

Warning

Synonyms: m-Anisylamine, 3-(Aminomethyl)anisole

159891-1G 159891-5G
220038-774EA 56.43 USD
220038-774 220038-775
3-Methoxybenzylamine ≥98%
3-Methoxybenzylamine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 238 °C (1013 hPa)
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00008115
CAS Number: 5071-96-5
EINECS: 225-779-6
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR