2-Methoxybenzylamine ≥98%

Danger

Synonyms: (2-Methoxyphenyl)methanamine, 2-(Aminomethyl)anisole, o-Anisylamine

159883-5G 159883-25G
220038-773EA 42.54 USD
220038-773 220038-772
2-Methoxybenzylamine ≥98%
2-Methoxybenzylamine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 227 °C (1013 hPa)
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008110
CAS Number: 6850-57-3
EINECS: 229-941-7
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR