Toluene-α-sulfonyl chloride ≥98%

Danger

Synonyms: Benzylsulphonyl chloride, α-Toluenesulphonyl chloride, Phenylmethylsulphonyl chloride, Phenylmethanesulphonyl chloride

159719-25G 159719-5G
220038-646EA 110.18 USD
220038-646 220039-052
Toluene-α-sulfonyl chloride ≥98%
Toluene-α-sulphonyl chloride
Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 120 °C (10 mmHg)
Melting Pt: 90…93 °C
MDL Number: MFCD00007455
CAS Number: 1939-99-7
EINECS: 217-717-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR