2,4-Dimethoxybenzyl alcohol ≥99%

159638-100G 159638-25G
220038-641EA 328.46 USD
220038-641 220038-642
2,4-Dimethoxybenzyl alcohol ≥99%
2,4-Dimethoxybenzyl alcohol
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 317 °C (1013 hPa)
Melting Pt: 34…35 °C
MDL Number: MFCD00004614
CAS Number: 7314-44-5
EINECS: 230-775-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR