Methyl-3,5-dihydroxybenzoate ≥97%

Synonyms: Methyl 3,5-dihydroxybenzoate, 3,5-Dihydroxybenzoic acid methyl ester

159603-25G 159603-100G
220038-638EA 59.77 USD
220038-638 220038-637
Methyl-3,5-dihydroxybenzoate ≥97%
Methyl-3,5-dihydroxybenzoate
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 168 - 170 °C
MDL Number: MFCD00002289
CAS Number: 2150-44-9
EINECS: 218-426-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR