6-Chloro-m-cresol ≥99%

Warning

Synonyms: 1-Chloro-2-hydroxy-4-methylbenzene, 2-Chloro-5-methylphenol

159557-25G 159557-100G
220038-632EA 87.02 USD
220038-632 220038-631
6-Chloro-m-cresol ≥99%
6-Chloro-m-cresol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 43…46 °C
Density: 1.215 g/cm³ (20 °C)
MDL Number: MFCD00002175
CAS Number: 615-74-7
EINECS: 210-444-9
UN: 3437
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR