ω-Bromo-4-methylacetophenone ≥90%

Synonyms: α-Bromo-4-methylacetophenone, Bromomethyl p-methylphenyl ketone, 2-Bromo-4'-methylacetophenone, 2-Bromo-1-(p-tolyl)ethanone, 4-Methylphenacyl bromide

159530-5G 159530-25G 159530-100G
220038-627EA 40.92 USD
220038-627 220038-626 220038-625
ω-Bromo-4-methylacetophenone ≥90%
ω-Bromo-4-methylacetophenone
Formula: C₉H₉BrO
MW: 213.07 g/mol
Boiling Pt: 105 °C (0.1 mmHg)
Melting Pt: 95…97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000203
CAS Number: 619-41-0
EINECS: 210-595-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR