12-Aminolauric acid ≥95%

159247-5G 159247-25G
220039-032EA 45.91 USD
220039-032 220039-031
12-Aminolauric acid ≥95%
12-Aminolauric acid
Formula: C₁₂H₂₅NO₂
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 183 °C
MDL Number: MFCD00008153
CAS Number: 693-57-2
EINECS: 211-754-7

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR