2-Bromoanisole ≥97%

Synonyms: 1-Bromo-2-methoxybenzene

159239-100G 159239-25G
220039-029EA 53.85 USD
220039-029 220039-030
2-Bromoanisole ≥97%
2-Bromoanisole
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 223 °C (1013 hPa)
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000064
CAS Number: 578-57-4
EINECS: 209-425-8
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR