Diethyl pyrocarbonate ≥97%

Warning

Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC

159220-100G 159220-5G 159220-25G
220039-026EA 304.77 USD
220039-026 220039-028 220039-027
Diethyl pyrocarbonate ≥97%
Diethyl pyrocarbonate
Formula: O(COOC₂H₅)₂
MW: 162.14 g/mol
Boiling Pt: 93 °C (1013 hPa)
Density: 1.123 g/cm³ (20 °C)
Flash Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR