4-Chloroanisole ≥99%

Synonyms: 1-Chloro-4-methoxybenzene

159069-100G 159069-25G
220039-016EA 308 USD
220039-016 220039-017
4-Chloroanisole ≥99%
4-Chloroanisole
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: -18 °C
Density: 1.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000616
CAS Number: 623-12-1
EINECS: 210-772-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR