Methyl toluene-4-sulfonate ≥98%

Warning

Synonyms: Methyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid methyl ester, Methyl p-toluenesulphonate, Toluene-4-sulphonic acid methyl ester

158992-500G 158992-100G
220039-005EA 103.82 USD
220039-005 220039-004
Methyl toluene-4-sulfonate ≥98%
Methyl toluene-4-sulphonate
Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 28…30 °C
Density: 1.23 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00008417
CAS Number: 80-48-8
EINECS: 201-283-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR