4'-Chloroacetanilide ≥97%

Synonyms: 4-Chloroacetanilide, Paracetamol impurity J, N-(4-Chlorophenyl)acetamide

158631-5G 158631-25G
220038-421EA 45.02 USD
220038-421 220038-420
4'-Chloroacetanilide ≥97%
4'-Chloroacetanilide
Formula: C₈H₈ClNO
MW: 169.61 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 179…180 °C
Density: 1.385 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000612
CAS Number: 539-03-7
EINECS: 208-707-8
Merck Index: 13,02128

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR