Methyl bromoacetate ≥97%

Synonyms: Bromoacetic methyl ester

157910-25G 157910-100G
220038-152EA 33.38 USD
220038-152 220038-151
Methyl bromoacetate ≥97%
Methyl bromoacetate
Formula: BrCH₂COOCH₃
MW: 152.98 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: -50 °C
Density: 1.635 g/cm³ (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00000189
CAS Number: 96-32-2
EINECS: 202-499-2
UN: 2643
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR