1,3-Dithiane

157872-5G 157872-25G
220038-147EA 56.54 USD
220038-147 220038-146
1,3-Dithiane
1,3-Dithiane
Formula: C₄H₈S₂
MW: 120.24 g/mol
Boiling Pt: 207 - 208 °C (1013 hPa)
Melting Pt: 51 - 54 °C
Density: 1.36 g/cm³ (25 °C)
MDL Number: MFCD00006654
CAS Number: 505-23-7
EINECS: 208-006-7
UN: 3335
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR