Phosphorus(III) bromide ≥97%

Danger

157783-100G 157783-500G
220038-187EA 108.77 USD
220038-187 220038-188
Phosphorus(III) bromide ≥97%
Phosphorus(III) bromide
Formula: PBr₃
Boiling Pt: 175 °C (1013 hPa)
Melting Pt: –40 °C
Density: 2.8903 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2
UN: 1808
ADR: 8,II
Merck Index: 12,07514

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR