2-Methylbenzophenone

Synonyms: o-Methylbenzophenone

157538-5G 157538-25G
220038-756EA 409.62 USD
220038-756 220038-755
2-Methylbenzophenone
2-Methylbenzophenone
Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.1098 g/cm³ (20 °C)
MDL Number: MFCD00008518
CAS Number: 131-58-8
EINECS: 205-032-0
Merck Index: 12,07470

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR