4-tert-Butylbenzoyl chloride ≥98%

Danger

Synonyms: 4-tert-Butylbenzoylchloride

157120-100G 157120-25G
220038-702EA 233.69 USD
220038-702 220038-703
4-tert-Butylbenzoyl chloride ≥98%
4-tert-Butylbenzoyl chloride
Formula: C₁₁H₁₃ClO
MW: 196.68 g/mol
Boiling Pt: 135 °C (1013 hPa)
Density: 1.0864 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000695
CAS Number: 1710-98-1
EINECS: 216-973-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR