2,6-Dimethylbenzoic acid ≥97%

156906-5G 156906-10G
220038-617EA 60 USD
220038-617 220038-616
2,6-Dimethylbenzoic acid ≥97%
2,6-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 155…160 °C (17 mmHg)
Melting Pt: 115…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002483
CAS Number: 632-46-2
EINECS: 211-177-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR