1-Hexyl-3-methylimidazolium chloride

Synonyms: 1-n-Hexyl-3-methylimidazolium chloride

15689-25G 15689-5G
220038-612EA 41.08 USD
220038-612 220038-613
1-Hexyl-3-methylimidazolium chloride
1-Hexyl-3-methylimidazolium chloride
Formula: C₁₀H₁₉ClN₂
MW: 202.73 g/mol
Melting Pt: –85 °C
Density: 1.05 g/cm³ (20 °C)
MDL Number: MFCD03093289
CAS Number: 171058-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR