L(+)-Diethyl tartrate ≥99%

Synonyms: (+)-Diethyl L-tartrate, (R,R)-(+)-Tartaric acid diethyl ester, (2R,3R)-(+)-Diethyl tartrate, Diethyl L-tartrate, L(+)-Tartaric acid diethyl ester, (2R,3R)-Diethyl 2,3-dihydroxysuccinate, (R,R)-(+)-Diethyl tartrate

156841-100G 156841-500G 156841-25G
220037-956EA 78.18 USD
220037-956 220037-958 220037-957
L(+)-Diethyl tartrate ≥99%
L(+)-Diethyl tartrate
Formula: C₈H₁₄O₆
MW: 206.2 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 18.7 °C
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD00009143
CAS Number: 87-91-2
EINECS: 201-783-3
Merck Index: 13,03886

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR