3',4'-Dimethoxyacetophenone ≥98%

Synonyms: 1-(3,4-Dimethoxyphenyl)ethan-1-one, 3,4-Dimethoxyacetophenone, 1-(3,4-Dimethoxyphenyl)ethanone

156639-100G 156639-25G
220038-398EA 205.69 USD
220038-398 220038-399
3',4'-Dimethoxyacetophenone ≥98%
3',4'-Dimethoxyacetophenone
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 48…49 °C
MDL Number: MFCD00008737
CAS Number: 1131-62-0
EINECS: 214-468-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR