1-Bromo-2-methylpropane ≥99%

Danger

Synonyms: Isobutyl bromide

156582-100G 156582-500G
220038-392EA 66.98 USD
220038-392 220038-393
1-Bromo-2-methylpropane ≥99%
1-Bromo-2-methylpropane
Formula: C₄H₉Br
MW: 137.02 g/mol
Boiling Pt: 91 °C (1013 hPa)
Melting Pt: –118 °C
Density: 1.265 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000217
CAS Number: 78-77-3
EINECS: 201-141-2
UN: 2342
ADR: 3,II
Merck Index: 13,05152

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR