2,4,5-Trichlorophenol ≥95%

Warning

156515-100G 156515-25G
220038-387EA 270.31 USD
220038-387 220038-388
2,4,5-Trichlorophenol ≥95%
2,4,5-Trichlorophenol
Formula: C₆H₃Cl₃O
MW: 197.45 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 68 °C
Density: 1.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002170
CAS Number: 95-95-4
EINECS: 202-467-8
UN: 3077
ADR: 9,III
Merck Index: 12,09772

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR