Cyclobutanol ≥99%

156434-1G 156434-5G
220038-588EA 150.77 USD
220038-588 220038-589
Cyclobutanol ≥99%
Cyclobutanol
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 122 °C (1013 hPa)
Density: 0.9226 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001318
CAS Number: 2919-23-5
EINECS: 220-858-1
UN: 1120
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR