2,3-Dimethyl-2,3-dinitrobutane ≥98%

Danger

156345-50G 156345-10G
220038-580EA 356.46 USD
220038-580 220038-579
2,3-Dimethyl-2,3-dinitrobutane ≥98%
2,3-Dimethyl-2,3-dinitrobutane
Formula: C₆H₁₂N₂O₄
MW: 176.17 g/mol
Melting Pt: 221 °C
Density: 1.34 g/cm³ (20 °C)
MDL Number: MFCD00007392
CAS Number: 3964-18-9
EINECS: 223-569-9
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR