Polyethylene glycol 6,000

81260-5KG 81260-1KG
220096-195EA 470.77 USD
220096-195 220097-070
Polyethylene glycol 6,000
Polyethylene glycol 6,000
Formula: H(OCH₂CH₂)nOH
Boiling Pt: ? 250 °C (1013 hPa)
Melting Pt: 55 - 62 °C
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779614
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR