Sodium methoxide 25% (w/w) in methanol

Danger

Synonyms: Sodium methylate, Sodium methanolate

156256-18L 156256-25ML 156256-1L
220038-181EA 366.92 USD
220038-181 220038-183 220038-182
Sodium methoxide 25% (w/w) in methanol
Sodium methoxide
Formula: CH₃NaO
MW: 54.02 g/mol
Density: 0.97 g/cm³ (20 °C)
MDL Number: MFCD00012179
CAS Number: 124-41-4
EINECS: 204-699-5
UN: 1289
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR