Sodium ethoxide ≥95%

Danger

Synonyms: Sodium ethylate, Sodium ethanolate

156248-100G 156248-5G 156248-500G
220038-178EA 51.69 USD
220038-178 220038-180 220038-179
Sodium ethoxide ≥95%
Sodium ethoxide
Formula: C₂H₅NaO
MW: 68.05 g/mol
Melting Pt: 260 °C
Density: 0.868 g/cm³ (20 °C)
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3206
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR