3-Hydroxy-4-nitrobenzaldehyde ≥97%

Warning

156167-10G 156167-1G
220038-171EA 300.46 USD
220038-171 220038-172
3-Hydroxy-4-nitrobenzaldehyde ≥97%
3-Hydroxy-4-nitrobenzaldehyde
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007109
CAS Number: 704-13-2
EINECS: 211-879-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR