2,4-Pentanediol ≥98%

Synonyms: 2,4-Dihydroxy pentane

156019-10G 156019-50G
220038-140EA 101.66 USD
220038-140 220038-141
2,4-Pentanediol ≥98%
2,4-Pentanediol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: ∼53 °C
Density: 0.96 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004549
CAS Number: 625-69-4
EINECS: 210-907-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR