4-Fluoro-3-nitroaniline ≥97%

Warning

155861-100G 155861-25G
220038-371EA 143.23 USD
220038-371 220038-372
4-Fluoro-3-nitroaniline ≥97%
4-Fluoro-3-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 96 - 98 °C
Density: 0.43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007833
CAS Number: 364-76-1
EINECS: 206-665-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR