Tetrabutylammonium hydrogen sulfate

Warning

Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate

155837-1KG 155837-25G 155837-100G
220038-367EA 296.15 USD
220038-367 220038-368 220038-366
Tetrabutylammonium hydrogen sulfate
Tetrabutylammonium hydrogen sulphate
Formula: (C₄H₉)₄NHSO₄
MW: 339.54 g/mol
Melting Pt: 169…172 °C
Density: 1.01 g/cm³ (25 °C)
MDL Number: MFCD00011637
CAS Number: 32503-27-8
EINECS: 251-068-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR