4-Fluorostyrene ≥99%

Synonyms: 1-Fluoro-4-vinylbenzene

155799-10G 155799-1G
220038-358EA 54.8 USD
220038-358 220038-359
4-Fluorostyrene ≥99%
4-Fluorostyrene
Formula: C₈H₇F
MW: 122.14 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –34.5 °C
Density: 1.022 g/cm³ (20 °C)
MDL Number: MFCD00000361
CAS Number: 405-99-2
EINECS: 206-975-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR