2,2,6,6-Tetramethyl-3,5-heptanedione ≥98%

Synonyms: Dipivaloylmethane, TMHD, 2,2,6,6-Tetramethylheptane-3,5-dione

155756-5G 155756-100G 155756-25G
220038-356EA 51.58 USD
220038-356 220038-354 220038-355
2,2,6,6-Tetramethyl-3,5-heptanedione ≥98%
2,2,6,6-Tetramethyl-3,5-heptanedione
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 28…30 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008848
CAS Number: 1118-71-4
EINECS: 214-268-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR