Methacrylic acid ≥99%

Danger

Synonyms: 2-Methylpropenoic acid

155721-100G 155721-2KG 155721-5G 155721-18KG 155721-3KG 155721-500G
220038-348EA 55.18 USD
220038-348 220038-350 220038-353 220038-349 220038-351 220038-352
Methacrylic acid ≥99%
Methacrylic acid
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: 15 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00002651
CAS Number: 79-41-4
EINECS: 201-204-4
UN: 2531
ADR: 8,II
Merck Index: 13,05967

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR