Polyethylene glycol 1,500

Warning

Synonyms: PEG 1500, PEG 1,500

81210-5KG 81210-1KG
220097-045EA 555.77 USD
220097-045 220097-044
Polyethylene glycol 1,500
Polyethylene glycol 1,500
Formula: H(OCH₂CH₂)nOH
MW: 1500 g/mol
Boiling Pt: ? 250 °C (1013 hPa)
Melting Pt: 44 - 48 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR