N,O-Bis(trimethylsilyl)trifluoroacetamide ≥99%

Danger

Synonyms: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA

155195-5G 155195-100G 155195-25G
220038-309EA 65.58 USD
220038-309 220038-307 220038-308
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥99%
N,O-Bis(trimethylsilyl)trifluoroacetamide
Formula: C₈H₁₈F₃NOSi₂
MW: 257.4 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –41 °C
Density: 0.96 g/cm³ (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00008269
CAS Number: 25561-30-2
EINECS: 247-103-9
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR