N(α)-Boc-L-tryptophan ≥99% (by TLC), Sigma-Aldrich®

Danger

Synonyms: Boc-Trp-OH , Boc-Trp-OH

15512-6X1L-R 15512-2.5L-R 15512-5L-R 15512-5G-F 15512-1L-R 15512-4X2.5L-R
220038-291EA 786.15 USD
220038-291 220038-286 220038-290 220038-289 220038-285 220038-287
N(α)-Boc-L-tryptophan ≥99% (by TLC), Sigma-Aldrich®
N(α)-Boc-L-tryptophan
Formula: C₁₆H₂₀N₂O₄
MW: 304.3 g/mol
Melting Pt: Max. -15
Storage Temperature: Ambient
MDL Number: MFCD00065595
CAS Number: 13139-14-5
EINECS: 200-001-8
ADR: 8,III

Order Now

Specification Test Results

Assay 99%
(TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR