4-Acetylbenzonitrile ≥99%

154393-25G 154393-5G
220037-245EA 621.38 USD
220037-245 220037-246
4-Acetylbenzonitrile ≥99%
4-Acetylbenzonitrile
Formula: C₉H₇NO
Boiling Pt: 110…112 °C (1 mmHg)
Melting Pt: 57…58 °C
MDL Number: MFCD00001825
CAS Number: 1443-80-7
EINECS: 215-885-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR