2-Thiophenecarbonitrile

Synonyms: 2-Thienyl cyanide, Thiophene-2-carbonitrile, 2-Cyanothiophene

154385-50G 154385-10G
220038-194EA 342.46 USD
220038-194 220038-138
2-Thiophenecarbonitrile
2-Thiophenecarbonitrile
Formula: C₅H₃NS
MW: 109.15 g/mol
Boiling Pt: 190 - 192 °C (1013 hPa)
Density: 1.18 g/cm³ (15 °C)
MDL Number: MFCD00005416
CAS Number: 1003-31-2
EINECS: 213-706-0
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR