4-Bromo-2-chloroaniline ≥98%

Warning

154245-10G 154245-50G
220037-162EA 132 USD
220037-162 220037-163
4-Bromo-2-chloroaniline ≥98%
4-Bromo-2-chloroaniline
Formula: C₆H₅BrClN
MW: 206.47 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00007660
CAS Number: 38762-41-3
EINECS: 254-118-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR