(R)-(-)-Mandelic acid ≥99%

Danger

Synonyms: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid

154210-5G 154210-25G
220037-158EA 39.85 USD
220037-158 220038-119
(R)-(-)-Mandelic acid ≥99%
(R)-(-)-Mandelic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 130 - 132 °C
Density: 1.341 g/cm³ (20 °C)
Flash Pt: ≥ 190 °C
MDL Number: MFCD00064251
CAS Number: 611-71-2
EINECS: 210-276-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR